8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
8-Chloro-6-(trifluoromethyl)imidazo-[1,2-a]pyridine-2-carboxylic acid
A680262-1G
A680262-250MG
A680262-5G
77158-216EA
20
USD
77158-216
77158-218
77158-220
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid 95%
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₄ClF₃N₂O₂ |
MDL Number:
MFCD00831510 CAS Number: 353258-35-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...