1-O-Methyl-2-deoxy-D-ribose 95%

Supplier: Ambeed
Warning

A678984-100G A678984-250MG A678984-25G
77158-210EA 292 USD
77158-210 77158-212 77158-214
1-O-Methyl-2-deoxy-D-ribose 95%
1-O-Methyl-2-deoxy-D-ribose

Precaución: for Research Use Only

Formula: C₆H₁₂O₄
MDL Number: MFCD00134161
CAS Number: 60134-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR