3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl 98%

Supplier: Ambeed
Warning

A676515-5G A676515-100G A676515-25G
77158-198EA 16 USD
77158-198 77900-652 77901-890
3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl 98%
3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl

Precaución: for Research Use Only

Formula: C₂₀H₂₂F₂
MDL Number: MFCD12828121
CAS Number: 134412-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR