Methyl 2-amino-6-methylnicotinate 95%

Supplier: Ambeed
Warning

A675776-100MG A675776-1G A675776-250MG A675776-5G
77158-192EA 44 USD
77158-192 77158-194 77158-196 77893-360
Methyl 2-amino-6-methylnicotinate 95%
Methyl 2-amino-6-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD16611580
CAS Number: 31686-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR