Cyclopentyl-2-pyridyl ketone 98%

Supplier: Ambeed
Warning

Synonyms: Cyclopentyl 2-pyridyl ketone, 2-Cyclopentanecarbonylpyridine

A675664-1G A675664-5G
77158-188EA 77.25 USD
77158-188 77158-190
Cyclopentyl-2-pyridyl ketone 98%
Cyclopentyl-2-pyridyl ketone

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
MDL Number: MFCD07699202
CAS Number: 157592-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR