N5-(3-(9H-Purin-6-yl)pyridin-2-yl)-N1-(4-chlorophenyl)-6-methylisoquinoline-1,5-diamine dihydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: Raf inhibitor 1 2HCl

A674211-10MG A674211-1MG A674211-25MG A674211-5MG A674211-50MG
77158-174EA 56.65 USD
77158-174 77158-176 77158-178 77158-180 77686-638
N5-(3-(9H-Purin-6-yl)pyridin-2-yl)-N1-(4-chlorophenyl)-6-methylisoquinoline-1,5-diamine dihydrochloride 98+%
N5-(3-(9H-Purin-6-yl)pyridin-2-yl)-N1-(4-chlorophenyl)-6-methylisoquinoline-1,5-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₂₆H₂₁Cl₃N₈
MW: 551.86 g/mol
MDL Number: MFCD18782676
CAS Number: 1191385-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR