2,4-Dichloro-5-nitrophenol 98%

Supplier: Ambeed
Danger

A673556-1G A673556-25G A673556-5G
77158-166EA 12 USD
77158-166 77158-168 77158-170
2,4-Dichloro-5-nitrophenol 98%
2,4-Dichloro-5-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₃
MDL Number: MFCD03093072
CAS Number: 39489-77-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR