N-(3-Fluoro-4-((7-(2-hydroxy-2-methylpropoxy)quinolin-4-yl)oxy)phenyl)-1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazole-4-carboxamide p-toluenesulfonate 95%

Supplier: Ambeed
Warning

A671711-10MG A671711-1MG A671711-25MG A671711-5MG
77158-160EA 36 USD
77158-160 77158-162 77158-164 77614-078
N-(3-Fluoro-4-((7-(2-hydroxy-2-methylpropoxy)quinolin-4-yl)oxy)phenyl)-1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazole-4-carboxamide p-toluenesulfonate 95%
N-(3-Fluoro-4-((7-(2-hydroxy-2-methylpropoxy)quinolin-4-yl)oxy)phenyl)-1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazole-4-carboxamide p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₃₈H₃₇FN₄O₈S
MDL Number: MFCD28142839
CAS Number: 1394820-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR