Methyl 5-chloro-1H-pyrazole-3-carboxylate 95%

Supplier: Ambeed
Warning

A669858-250MG A669858-5G A669858-1G
77158-144EA 23 USD
77158-144 77158-146 77893-352
Methyl 5-chloro-1H-pyrazole-3-carboxylate 95%
Methyl 5-chloro-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂
MDL Number: MFCD29037474
CAS Number: 1810069-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR