2-(Tosyloxy)acetic acid 95%

Supplier: Ambeed
Warning

A667600-100MG A667600-250MG A667600-1G
77158-130EA 18 USD
77158-130 77158-132 77573-150
2-(Tosyloxy)acetic acid 95%
2-(Tosyloxy)acetic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₅S
MDL Number: MFCD09892424
CAS Number: 39794-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR