Isoamyl laurate 98+%

Supplier: Ambeed
Warning

A667325-100G A667325-500G
77158-126EA 6 USD
77158-126 77158-128
Isoamyl laurate 98+%
Isoamyl laurate

Precaución: for Research Use Only

Formula: C₁₇H₃₄O₂
MDL Number: MFCD00048429
CAS Number: 6309-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR