Hexaammineruthenium trichloride ≥98% Ru ≥32.1%

Supplier: Ambeed
Warning

Synonyms: Hexaammineruthenium(III) chloride, Hexaammineruthenium (III) chloride

A665970-5G A665970-250MG
77158-118EA 169 USD
77158-118 77614-052
Hexaammineruthenium trichloride ≥98% Ru ≥32.1%
Hexaammineruthenium trichloride

Precaución: for Research Use Only

Formula: H₁₈Cl₃N₆Ru
MW: 309.61 g/mol
MDL Number: MFCD00011478
CAS Number: 14282-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR