4-(Difluoromethoxy)iodobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Difluoromethoxy)-4-iodobenzene

A665698-100G A665698-1G A665698-25G A665698-5G
77158-110EA 201.88 USD
77158-110 77158-112 77158-114 77158-116
4-(Difluoromethoxy)iodobenzene 98%
4-(Difluoromethoxy)iodobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂IO
MW: 270.0153 g/mol
MDL Number: MFCD00085008
CAS Number: 128140-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR