Methyl (4,6-diamino-2-(5-fluoro-1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl)pyrimidin-5-yl)carbamate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Vericiguat
A664188-50MG
A664188-1G
77158-092EA
160.68
USD
77158-092
77686-631
Methyl (4,6-diamino-2-(5-fluoro-1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl)pyrimidin-5-yl)carbamate 99%
Methyl (4,6-diamino-2-(5-fluoro-1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl)pyrimidin-5-yl)carbamate
Precaución: for Research Use Only
Formula:
C₁₉H₁₆F₂N₈O₂ MW: 426.38 g/mol |
MDL Number:
MFCD28502029 CAS Number: 1350653-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...