(S)-1-Benzyl 3-methyl piperidine-1,3-dicarboxylate 98+%
Supplier: Ambeed
|
Warning
|
A662530-100MG
A662530-250MG
77158-084EA
145.23
USD
77158-084
77158-086
(S)-1-Benzyl 3-methyl piperidine-1,3-dicarboxylate 98+%
(S)-1-Benzyl 3-methyl piperidine-1,3-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₁₉NO₄ MW: 277.31 g/mol |
MDL Number:
MFCD17168402 CAS Number: 88466-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...