(3-Nitropyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Hydroxymethyl)-3-nitropyridine

A661975-100MG A661975-250MG
77158-074EA 111.24 USD
77158-074 77158-076
(3-Nitropyridin-2-yl)methanol 97%
(3-Nitropyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD18802584
CAS Number: 36625-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR