N,N-Dimethylglycine ethyl ester 98%

Supplier: Ambeed
Danger

Synonyms: Di-Me-Gly-OEt

A660129-25G A660129-5G
77158-056EA 49.44 USD
77158-056 77158-058
N,N-Dimethylglycine ethyl ester 98%
N,N-Dimethylglycine ethyl ester

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
MDL Number: MFCD00009175
CAS Number: 33229-89-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR