5-Chlorothiazole-2-ethanone 95%

Supplier: Ambeed
Warning

A659567-1G A659567-250MG A659567-5G
77158-050EA 115.36 USD
77158-050 77158-052 77158-054
5-Chlorothiazole-2-ethanone 95%
5-Chlorothiazole-2-ethanone

Precaución: for Research Use Only

Formula: C₅H₄ClNOS
MW: 161.60 g/mol
MDL Number: MFCD09842149
CAS Number: 774230-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR