3-(4-Bromophenyl)oxetane 95%

Supplier: Ambeed
Warning

A659092-100MG A659092-1G A659092-250MG A659092-50MG
77158-042EA 59 USD
77158-042 77158-044 77158-046 77158-048
3-(4-Bromophenyl)oxetane 95%
3-(4-Bromophenyl)oxetane

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD22628728
CAS Number: 1402158-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR