Y-320 98%

Supplier: Ambeed
Warning

Synonyms: 1-​(4-​chlorophenyl)​-​N-​[3-​cyano-​4-​[4-​(4-​morpholinyl)​-​1-​piperidinyl]​phenyl]​-​5-​methyl-1H-​pyrazole-​4-​carboxamide, 1-(4-Chlorophenyl)-N-(3-cyano-4-(4-morpholinopiperidin-1-yl)phenyl)-5-methyl-1H-pyrazole-4-carboxamide

A657468-10MG A657468-1MG A657468-50MG A657468-5MG
77158-034EA 83.43 USD
77158-034 77158-036 77158-038 77158-040
Y-320 98%
Y-320

Precaución: for Research Use Only

Formula: C₂₇H₂₉ClN₆O₂
MW: 505.01 g/mol
MDL Number: MFCD28099803
CAS Number: 288250-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR