1-Hexyl-3-methylimidazolium trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

A656906-100G A656906-25G
77158-030EA 120 USD
77158-030 77158-032
1-Hexyl-3-methylimidazolium trifluoromethanesulfonate 98%
1-Hexyl-3-methylimidazolium trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₁₁H₁₉F₃N₂O₃S
MDL Number: MFCD03427621
CAS Number: 460345-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR