Ethyl 4-(cyclohexylamino)-3-nitrobenzoate 98%

Supplier: Ambeed
Warning

A655246-1G A655246-5G
77158-018EA 24.72 USD
77158-018 77158-020
Ethyl 4-(cyclohexylamino)-3-nitrobenzoate 98%
Ethyl 4-(cyclohexylamino)-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₂₀N₂O₄
MW: 292.33 g/mol
MDL Number: MFCD06016735
CAS Number: 87815-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR