1-(Isopropylsulfonyl)piperazine 95%

Supplier: Ambeed
Warning

A650988-100MG A650988-250MG
77158-006EA 46.35 USD
77158-006 77158-008
1-(Isopropylsulfonyl)piperazine 95%
1-(Isopropylsulfonyl)piperazine

Precaución: for Research Use Only

Formula: C₇H₁₆N₂O₂S
MW: 192.27 g/mol
MDL Number: MFCD09743769
CAS Number: 534615-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR