2-(Diethylamino)ethyl bromide hydrobromide 95%

Supplier: Ambeed
Danger

Synonyms: 2-Bromotriethylamine Hydrobromide, 2-Bromoethyldiethylamine Hydrobromide, 2-Bromo-N,N-diethylethylamine Hydrobromide

A649335-1G A649335-25G A649335-5G
77158-000EA 6.18 USD
77158-000 77158-002 77158-004
2-(Diethylamino)ethyl bromide hydrobromide 95%
2-(Diethylamino)ethyl bromide hydrobromide

Precaución: for Research Use Only

Formula: C₆H₁₄BrN
MW: 261.00 g/mol
MDL Number: MFCD00040376
CAS Number: 1069-72-3
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR