(S)-5-(Hydroxymethyl)oxazolidin-2-one 95%

Supplier: Ambeed
Warning

A647419-100MG A647419-1G A647419-250MG
77157-986EA 100.94 USD
77157-986 77157-988 77157-990
(S)-5-(Hydroxymethyl)oxazolidin-2-one 95%
(S)-5-(Hydroxymethyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₄H₇NO₃
MW: 117.1 g/mol
MDL Number: MFCD27977273
CAS Number: 97859-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR