7-(tert-Butoxycarbonyl)-2-oxa-7-azaspiro-[4.5]decane-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A645093-100MG
A645093-1G
77157-982EA
194
USD
77157-982
77157-984
7-(tert-Butoxycarbonyl)-2-oxa-7-azaspiro-[4.5]decane-3-carboxylic acid 97%
7-(tert-Butoxycarbonyl)-2-oxa-7-azaspiro-[4.5]decane-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₂₃NO₅ |
MDL Number:
MFCD12198530 CAS Number: 1160246-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...