4'-Hydroxy-[1,1'-biphenyl]-4-carbaldehyde 98%

Supplier: Ambeed
Warning

A643784-100MG A643784-250MG A643784-1G
77157-978EA 51.5 USD
77157-978 77157-980 77686-595
4'-Hydroxy-[1,1'-biphenyl]-4-carbaldehyde 98%
4'-Hydroxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MW: 198.21 g/mol
MDL Number: MFCD00804656
CAS Number: 100980-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR