2,7-Bis[N-(1-naphthyl)anilino]-9,9'-spirobi[9H-fluorene] 98+%

Supplier: Ambeed
Warning

A643037-100MG A643037-1G A643037-250MG A643037-5G
77157-970EA 10 USD
77157-970 77157-972 77157-974 77157-976
2,7-Bis[N-(1-naphthyl)anilino]-9,9'-spirobi[9H-fluorene] 98+%
2,7-Bis[N-(1-naphthyl)anilino]-9,9'-spirobi[9H-fluorene]

Precaución: for Research Use Only

Formula: C₅₇H₃₈N₂
MDL Number: MFCD12022428
CAS Number: 932739-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR