2,5-Difluorobenzenesulfonyl chloride 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,5-Difluorobenzene-1-sulphonyl chloride
A642197-10G
A642197-25G
A642197-5G
77157-960EA
10.3
USD
77157-960
77157-962
77157-964
2,5-Difluorobenzenesulfonyl chloride 97%
2,5-Difluorobenzenesulphonyl chloride
Precaución: for Research Use Only
Formula:
C₆H₃ClF₂O₂S MW: 212.6 g/mol |
MDL Number:
MFCD00060682 CAS Number: 26120-86-5 UN: 3265 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...