5-Bromo-2-furoyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 5-Bromofuran-2-carbonyl chloride

A641526-1G A641526-250MG
77157-950EA 145.23 USD
77157-950 77157-952
5-Bromo-2-furoyl chloride 95%
5-Bromo-2-furoyl chloride

Precaución: for Research Use Only

Formula: C₅H₂BrClO₂
MW: 209.43 g/mol
MDL Number: MFCD03424743
CAS Number: 26726-16-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR