tert-Butyl-3-aminobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-aminobenzoate

A640854-1G A640854-25G A640854-5G
77157-942EA 22.66 USD
77157-942 77157-944 77157-946
tert-Butyl-3-aminobenzoate 97%
tert-Butyl-3-aminobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MW: 193.2423 g/mol
MDL Number: MFCD00729048
CAS Number: 92146-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR