Balsalazide disodium salt dihydrate 98%

Supplier: Ambeed
Warning

Synonyms: 5-[4-(2-Carboxyethylcarbamoyl)phenylazo]salicylic Acid Disodium Salt, Sodium (E)-5-((4-((2-carboxylatoethyl)carbamoyl)phenyl)diazenyl)-2-hydroxybenzoate dihydrate

A639284-100MG A639284-10MG A639284-1MG A639284-50MG
77157-926EA 13.39 USD
77157-926 77157-928 77157-930 77157-932
Balsalazide disodium salt dihydrate 98%
Balsalazide disodium salt dihydrate

Precaución: for Research Use Only

Formula: C₁₇H₁₇N₃Na₂O₈
MW: 437.31 g/mol
MDL Number: MFCD01631117
CAS Number: 150399-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR