2,3-Difluorobenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Difluorophenylboronic acid pinacol ester, 1,3,2-Dioxaborolane, 2-(2,3-difluorophenyl)-4,4,5,5-tetramethyl-, 2-(2,3-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A638803-1G A638803-250MG A638803-5G
77157-914EA 41.2 USD
77157-914 77157-916 77157-918
2,3-Difluorobenzeneboronic acid pinacol ester 98%
2,3-Difluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BF₂O₂
MW: 240.05 g/mol
MDL Number: MFCD12405347
CAS Number: 1073339-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR