N-Methyl Omeprazole 95%

Supplier: Ambeed
Warning

A638530-100MG A638530-250MG
77157-910EA 190.55 USD
77157-910 77157-912
N-Methyl Omeprazole 95%
N-Methyl Omeprazole

Precaución: for Research Use Only

Formula: C₁₈H₂₁N₃O₃S
MW: 359.44 g/mol
MDL Number: MFCD19441777
CAS Number: 89352-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR