2-Bromo-4-fluorobenzenesulfonyl chloride 98%
Supplier: Ambeed
|
Danger
|
A634337-25G
A634337-5G
77157-890EA
53.56
USD
77157-890
77157-892
2-Bromo-4-fluorobenzenesulfonyl chloride 98%
2-Bromo-4-fluorobenzenesulphonyl chloride
Precaución: for Research Use Only
Formula:
C₆H₃BrClFO₂S MW: 273.51 g/mol |
MDL Number:
MFCD03094149 CAS Number: 351003-45-7 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...