(9H-Fluoren-9-yl)methyl (4-aminobutyl)carbamate hydrochloride 95%

Supplier: Ambeed
Warning

A634094-1G A634094-250MG A634094-5G
77157-882EA 66.95 USD
77157-882 77157-884 77157-886
(9H-Fluoren-9-yl)methyl (4-aminobutyl)carbamate hydrochloride 95%
(9H-Fluoren-9-yl)methyl (4-aminobutyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₂₃ClN₂O₂
MW: 346.85 g/mol
MDL Number: MFCD01631652
CAS Number: 321660-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR