N-(6-Bromonaphthalen-2-yl)methanesulfonamide 95%

Supplier: Ambeed
Warning

A633684-100MG A633684-1G A633684-250MG
77157-876EA 23 USD
77157-876 77157-878 77157-880
N-(6-Bromonaphthalen-2-yl)methanesulfonamide 95%
N-(6-Bromonaphthalen-2-yl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₂S
MDL Number: MFCD13191640
CAS Number: 1132940-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR