2-(4-Bromophenyl)ethanamine, N-BOC protected 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-2-(4-Bromo-phenyl)-ethylamine, N-Boc-2-(4-Bromophenyl)ethylamine

A631377-1G A631377-250MG A631377-10G A631377-5G A631377-25G A631377-100G
77157-870EA 8 USD
77157-870 77157-872 77613-952 77613-954 77686-575 77900-987
2-(4-Bromophenyl)ethanamine, N-BOC protected 97%
2-(4-Bromophenyl)ethanamine, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₂
MDL Number: MFCD08443684
CAS Number: 120157-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR