2-Bromo-4-fluoro-6-nitrophenol 98%

Supplier: Ambeed
Danger

A630809-1G A630809-250MG A630809-5G A630809-25G
77157-864EA 10 USD
77157-864 77157-866 77157-868 77900-612
2-Bromo-4-fluoro-6-nitrophenol 98%
2-Bromo-4-fluoro-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₃
MDL Number: MFCD03094182
CAS Number: 320-75-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR