A-803467 98%

Supplier: Ambeed
Warning

Synonyms: 5-(4-Chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide

A630517-10MG A630517-1MG A630517-50MG
77157-852EA 71.07 USD
77157-852 77157-854 77157-856
A-803467 98%
A-803467

Precaución: for Research Use Only

Formula: C₁₉H₁₆ClNO₄
MW: 357.79 g/mol
MDL Number: MFCD10574689
CAS Number: 944261-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR