SAR-20347 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Chloro-6-fluorophenyl)-5-((4-(morpholine-4-carbonyl)phenyl)amino)oxazole-4-carboxamide, 2-(2-Chloro-6-fluorophenyl)-5-[[4-(4-morpholinylcarbonyl)phenyl]amino]-4-oxazolecarboxamide

A627263-10MG A627263-1MG A627263-25MG A627263-5MG
77157-828EA 76.22 USD
77157-828 77157-830 77157-832 77157-834
SAR-20347 98%
SAR-20347

Precaución: for Research Use Only

Formula: C₂₁H₁₈ClFN₄O₄
MW: 444.84 g/mol
MDL Number: MFCD28502117
CAS Number: 1450881-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR