Dibutyl vinylboronate 94% stabilised

Supplier: Ambeed
Warning

Synonyms: Vinylboronic Acid Dibutyl Ester, Dibutyl Vinylboronate

Stabilization: Stabilised with phenothiazine

A626037-1G A626037-250MG
77157-812EA 20 USD
77157-812 77157-814
Dibutyl vinylboronate 94% stabilised
Dibutyl vinylboronate

Precaución: for Research Use Only

Formula: C₁₀H₂₁BO₂
MW: 184.09 g/mol
MDL Number: MFCD00050049
CAS Number: 6336-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR