4,4,5,5-Tetramethyl-2-(4-propoxyphenyl)-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A625227-1G A625227-250MG A625227-5G
77157-800EA 76.22 USD
77157-800 77157-802 77157-804
4,4,5,5-Tetramethyl-2-(4-propoxyphenyl)-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-(4-propoxyphenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₃
MW: 262.15 g/mol
MDL Number: MFCD22718113
CAS Number: 1374430-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR