1-Methyl-3-octylimidazolium trifluoromethansulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methyl-3-n-octylimidazolium trifluoromethanesulphonate, 1-Methyl-3-n-octylimidazolium triflate, 1-Methyl-3-n-octylimidazolium Trifluoromethanesulphonate
A623426-1G
A623426-25G
A623426-5G
77157-772EA
12.36
USD
77157-772
77157-774
77157-776
1-Methyl-3-octylimidazolium trifluoromethansulfonate 98%
1-Methyl-3-octylimidazolium trifluoromethansulphonate
Precaución: for Research Use Only
Formula:
C₁₃H₂₃F₃N₂O₃S MW: 344.39 g/mol |
MDL Number:
MFCD06411717 CAS Number: 403842-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...