4-((9-Cyclopentyl-7,7-difluoro-5-methyl-6-oxo-6,7,8,9-tetrahydro-5H-pyrimido[4,5-b][1,4]diazepin-2-yl)amino)-2-fluoro-5-methoxy-N-(1-methylpiperidin-4-yl)benzamide ≥98%

Supplier: Ambeed
Warning

Synonyms: TAK-960

A621588-10MG A621588-1MG A621588-5MG
77157-760EA 130.81 USD
77157-760 77157-762 77157-764
4-((9-Cyclopentyl-7,7-difluoro-5-methyl-6-oxo-6,7,8,9-tetrahydro-5H-pyrimido[4,5-b][1,4]diazepin-2-yl)amino)-2-fluoro-5-methoxy-N-(1-methylpiperidin-4-yl)benzamide ≥98%
4-((9-Cyclopentyl-7,7-difluoro-5-methyl-6-oxo-6,7,8,9-tetrahydro-5H-pyrimido[4,5-b][1,4]diazepin-2-yl)amino)-2-fluoro-5-methoxy-N-(1-methylpiperidin-4-yl)benzamide

Precaución: for Research Use Only

Formula: C₂₇H₃₄F₃N₇O₃
MW: 561.6 g/mol
MDL Number: MFCD22420821
CAS Number: 1137868-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR