(2S)-Isopropyl 2-(((perfluorophenoxy)(phenoxy)phosphoryl)amino)propanoate 95%

Supplier: Ambeed
Warning

A621578-10G A621578-1G A621578-25G
77157-754EA 166.86 USD
77157-754 77157-756 77157-758
(2S)-Isopropyl 2-(((perfluorophenoxy)(phenoxy)phosphoryl)amino)propanoate 95%
(2S)-Isopropyl 2-(((perfluorophenoxy)(phenoxy)phosphoryl)amino)propanoate

Precaución: for Research Use Only

Formula: C₁₈H₁₇F₅NO₅P
MW: 453.29 g/mol
MDL Number: MFCD22665799
CAS Number: 1256490-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR