tert-Butyl 4-methyl-5,6-dihydropyridine-1(2H)-carboxylate 98+%

Supplier: Ambeed
Danger

A620473-100MG A620473-1G A620473-250MG
77157-748EA 72.1 USD
77157-748 77157-750 77157-752
tert-Butyl 4-methyl-5,6-dihydropyridine-1(2H)-carboxylate 98+%
tert-Butyl 4-methyl-5,6-dihydropyridine-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₂
MW: 197.27 g/mol
MDL Number: MFCD26959125
CAS Number: 208245-73-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR