4,5-Dihydro-2-(methylthio)-1H-imidazole-1-carboxylic acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(methylthio)-4,5-dihydro-1H-imidazole-1-carboxylate

A620288-1G A620288-250MG
77157-744EA 124 USD
77157-744 77157-746
4,5-Dihydro-2-(methylthio)-1H-imidazole-1-carboxylic acid methyl ester 95%
4,5-Dihydro-2-(methylthio)-1H-imidazole-1-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O₂S
MDL Number: MFCD25965248
CAS Number: 60546-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR