N-(4-Chloro-3-(6-(dimethylamino)-1H-benzo[d]imidazol-2-yl)phenyl)-3,5-dimethoxybenzamide 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
HhAntag
A619601-10MG
A619601-1MG
A619601-25MG
A619601-50MG
A619601-5MG
77157-734EA
136.99
USD
77157-734
77157-736
77157-738
77157-740
77157-742
N-(4-Chloro-3-(6-(dimethylamino)-1H-benzo[d]imidazol-2-yl)phenyl)-3,5-dimethoxybenzamide 95%
N-(4-Chloro-3-(6-(dimethylamino)-1H-benzo[d]imidazol-2-yl)phenyl)-3,5-dimethoxybenzamide
Precaución: for Research Use Only
Formula:
C₂₄H₂₃ClN₄O₃ MW: 450.92 g/mol |
MDL Number:
MFCD25976715 CAS Number: 496794-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...