(R)-(-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine 98%

Supplier: Ambeed
Warning

A619420-100G A619420-1G A619420-25G A619420-5G
77157-726EA 904.01 USD
77157-726 77157-728 77157-730 77157-732
(R)-(-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine 98%
(R)-(-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD05864624
CAS Number: 122536-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR